α,α,α,2,4-Pentafluorotoluene ≥97%

Supplier: Matrix Scientific

071484-1G 071484-5G
102520-188EA 110.77 USD
102520-188 102520-190
α,α,α,2,4-Pentafluorotoluene ≥97%
α,α,α,2,4-Pentafluorotoluene

Hazard: Irritant

Formula: C₇H₃F₅
MDL Number: MFCD04972808
CAS Number: 64248-61-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR