3-Fluoro-4-methylbenzotrifluoride ≥95%
Supplier: Matrix Scientific
071465-1G
071465-5G
102520-144EA
207.69
USD
102520-144
102520-146
3-Fluoro-4-methylbenzotrifluoride ≥95%
3-Fluoro-4-methylbenzotrifluoride
Hazard: Irritant
Formula:
C₈H₆F₄ |
CAS Number:
1204296-09-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...