Pyrido[3,4-d]pyrimidin-4(3H)-one ≥97%

Supplier: Matrix Scientific

071446-500MG 071446-1G
102520-098EA 403.08 USD
102520-098 102520-100
Pyrido[3,4-d]pyrimidin-4(3H)-one ≥97%
Pyrido[3,4-d]pyrimidin-4(3H)-one

Hazard: Irritant

Formula: C7H5N3O
MW: 147.1300 g/mol
Melting Pt: 320 °C (dec.)
MDL Number: MFCD09999183
CAS Number: 19178-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR