2-Methyl-2-[(4-methylphenyl)amino]propanenitrile ≥97%

Supplier: Matrix Scientific

071443-500MG 071443-1G 071443-5G
102520-086EA 193.85 USD
102520-086 102520-088 102520-090
2-Methyl-2-[(4-methylphenyl)amino]propanenitrile ≥97%
2-Methyl-2-[(4-methylphenyl)amino]propanenitrile

Hazard: Irritant

Formula: C₁₁H₁₄N₂
Melting Pt: 77…80 °C
MDL Number: MFCD00719068
CAS Number: 101568-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR