Methyl-3-amino-5-bromo-4-hydroxybenzoate ≥97%

Supplier: Matrix Scientific

071441-500MG 071441-1G
102520-080EA 600 USD
102520-080 102520-082
Methyl-3-amino-5-bromo-4-hydroxybenzoate ≥97%
Methyl-3-amino-5-bromo-4-hydroxybenzoate

Hazard: Irritant

Formula: C₈H₈BrNO₃
MW: 246.07 g/mol
Melting Pt: 134…136 °C
MDL Number: MFCD22126120
CAS Number: 260249-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR