3-Iodo-1H-indole-2-carbonitrile ≥97%

Supplier: Matrix Scientific

071438-500MG 071438-1G
102520-068EA 515.38 USD
102520-068 102520-070
3-Iodo-1H-indole-2-carbonitrile ≥97%
3-Iodo-1H-indole-2-carbonitrile

Hazard: Irritant

Formula: C₉H₅IN₂
Melting Pt: 178…180 °C
MDL Number: MFCD22126121
CAS Number: 51796-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR