8-Aminoimidazo[1,2-a]pyridine ≥97%

Supplier: Matrix Scientific

Synonyms: Imidazo[1,2-a]pyridin-8-ylamine

071437-500MG 071437-1G
102520-064EA 130.77 USD
102520-064 102520-066
8-Aminoimidazo[1,2-a]pyridine ≥97%
8-Aminoimidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₇H₇N₃
MW: 133.1500 g/mol
Melting Pt: 40-43°C
MDL Number: MFCD06739233
CAS Number: 73221-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR