1-(4,6-Dichloropyridin-3-yl)ethan-1-one ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(4,6-Dichloropyridin-3-yl)ethanone, 5-Acetyl-2,4-dichloropyridine

071433-500MG 071433-1G
102520-050EA 615.38 USD
102520-050 102520-052
1-(4,6-Dichloropyridin-3-yl)ethan-1-one ≥95%
1-(4,6-Dichloropyridin-3-yl)ethan-1-one

Hazard: Irritant

Formula: C₇H₅Cl₂NO
MW: 190.03 g/mol
MDL Number: MFCD03265322
CAS Number: 887573-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR