2-Chloro-4-(trifluoromethyl)pyridine-3-carboxamide ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-4-(trifluoromethyl)nicotinamide

071429-500MG 071429-1G
102520-034EA 600 USD
102520-034 102520-036
2-Chloro-4-(trifluoromethyl)pyridine-3-carboxamide ≥97%
2-Chloro-4-(trifluoromethyl)pyridine-3-carboxamide

Hazard: Irritant

Formula: C₇H₄ClF₃N₂O
MW: 224.57 g/mol
Melting Pt: 187…188 °C
MDL Number: MFCD04039215
CAS Number: 886762-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR