3-Chloro-8-nitroimidazo[1,2-a]pyridine ≥97%

Supplier: Matrix Scientific

071426-500MG 071426-1G
102520-022EA 403.08 USD
102520-022 102520-024
3-Chloro-8-nitroimidazo[1,2-a]pyridine ≥97%
3-Chloro-8-nitroimidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₇H₄ClN₃O₂
Melting Pt: 172…174 °C
MDL Number: MFCD22126127
CAS Number: 52310-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR