3-Bromo-8-nitroimidazo[1,2-a]pyridine ≥97%

Supplier: Matrix Scientific

071423-500MG 071423-1G
102520-012EA 307.69 USD
102520-012 102520-014
3-Bromo-8-nitroimidazo[1,2-a]pyridine ≥97%
3-Bromo-8-nitroimidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₇H₄BrN₃O₂
Melting Pt: 198…201 °C
MDL Number: MFCD17016106
CAS Number: 52310-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR