Methyl-2-amino-6-fluoro-3-nitrobenzoate ≥98%

Supplier: Matrix Scientific

Synonyms: Methyl 2-amino-6-fluoro-3-nitrobenzoate

071414-1G 071414-5G
102519-980EA 89.23 USD
102519-980 102519-982
Methyl-2-amino-6-fluoro-3-nitrobenzoate ≥98%
Methyl-2-amino-6-fluoro-3-nitrobenzoate

Hazard: Irritant

Formula: C₈H₇FN₂O₄
MW: 214.15 g/mol
MDL Number: MFCD09839221
CAS Number: 346691-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR