4-Bromo-2-(difluoromethyl)-1-fluorobenzene ≥97%

Supplier: Matrix Scientific

071383-1G 071383-5G
102519-964EA 44.62 USD
102519-964 102519-966
4-Bromo-2-(difluoromethyl)-1-fluorobenzene ≥97%
4-Bromo-2-(difluoromethyl)-1-fluorobenzene

Hazard: Irritant

Formula: C₇H₄BrF₃
Boiling Pt: 188°C
Density: 1.633
Flash Pt: 74.9°C
MDL Number: MFCD06411633
CAS Number: 445303-69-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR