Trifluoromethyl-4-{1H-imidazo[4,5-b]pyridin-1-yl}piperidine-1-carboxylate ≥97%

Supplier: Matrix Scientific

071379-500MG 071379-1G
102519-958EA 803.08 USD
102519-958 102519-960
Trifluoromethyl-4-{1H-imidazo[4,5-b]pyridin-1-yl}piperidine-1-carboxylate ≥97%
Trifluoromethyl-4-{1H-imidazo[4,5-b]pyridin-1-yl}piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₃F₃N₄O₂
MW: 314.27 g/mol
MDL Number: MFCD22157002
CAS Number: 273757-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR