1-(Piperidin-4-yl)-1H,2H,3H-imidazo[4,5-b]pyridin-2-one dihydrochloride ≥97%
Supplier: Matrix Scientific
071374-500MG
071374-1G
102519-940EA
803.08
USD
102519-940
102519-942
1-(Piperidin-4-yl)-1H,2H,3H-imidazo[4,5-b]pyridin-2-one dihydrochloride ≥97%
1-(Piperidin-4-yl)-1H,2H,3H-imidazo[4,5-b]pyridin-2-one dihydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₆Cl₂N₄O Melting Pt: >350 °C |
MDL Number:
MFCD14705173 CAS Number: 781649-84-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...