5-Methylpyridine-2-carboxylic acid hydrochloride ≥95%
Supplier: Matrix Scientific
071369-1G
071369-5G
102519-922EA
76.92
USD
102519-922
102519-924
5-Methylpyridine-2-carboxylic acid hydrochloride ≥95%
5-Methylpyridine-2-carboxylic acid hydrochloride
Hazard: Irritant
Formula:
C₇H₈ClNO₂ Melting Pt: >300 °C |
MDL Number:
MFCD20040409 CAS Number: 177359-60-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...