5-(2-Methyloctan-2-yl)benzene-1,3-diol ≥95%

Supplier: Matrix Scientific

Synonyms: 5-(1,1-Dimethylheptyl)resorcinol, 5-(2-Methyloctan-2-yl)benzene-1,3-diol

071364-500MG 071364-1G
102519-902EA 403.08 USD
102519-902 102519-904
5-(2-Methyloctan-2-yl)benzene-1,3-diol ≥95%
5-(2-Methyloctan-2-yl)benzene-1,3-diol

Hazard: Irritant

Formula: C₁₅H₂₄O₂
MW: 236.36 g/mol
MDL Number: MFCD03001733
CAS Number: 56469-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR