Methyl-3-iodoimidazo[1,2-a]pyridine-7-carboxylate ≥97%

Supplier: Matrix Scientific

071360-500MG 071360-1G
102519-886EA 403.08 USD
102519-886 102519-888
Methyl-3-iodoimidazo[1,2-a]pyridine-7-carboxylate ≥97%
Methyl-3-iodoimidazo[1,2-a]pyridine-7-carboxylate

Hazard: Irritant

Formula: C₉H₇IN₂O₂
Melting Pt: 218…219 °C
MDL Number: MFCD12400833
CAS Number: 1009378-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR