{2-Methylimidazo[1,2-a]pyridin-3-yl}methanol ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(Hydroxymethyl)-2-methylimidazo[1,2-a]pyridine

071359-500MG 071359-1G
102519-882EA 403.08 USD
102519-882 102519-884
{2-Methylimidazo[1,2-a]pyridin-3-yl}methanol ≥95%
{2-Methylimidazo[1,2-a]pyridin-3-yl}methanol

Hazard: Irritant

Formula: C₉H₁₀N₂O
MW: 162.19 g/mol
Melting Pt: 158…160 °C
MDL Number: MFCD07368506
CAS Number: 30489-44-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR