imidazo[1,2-a]pyridine-7-carbohydrazide ≥95%

Supplier: Matrix Scientific

Synonyms: Imidazo[1,2-a]pyridine-7-carbohydrazide

071350-500MG 071350-1G
102519-846EA 403.08 USD
102519-846 102519-848
imidazo[1,2-a]pyridine-7-carbohydrazide ≥95%
imidazo[1,2-a]pyridine-7-carbohydrazide

Hazard: Irritant

Formula: C₈H₈N₄O
MW: 176.179 g/mol
Melting Pt: 269…270 °C
MDL Number: MFCD13183226
CAS Number: 421595-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR