{[2-Hydroxy-3-(phenylsulfanyl)propyl]sulfanyl}benzene ≥97%

Supplier: Matrix Scientific

071349-500MG 071349-1G 071349-5G
102519-840EA 193.85 USD
102519-840 102519-842 102519-844
{[2-Hydroxy-3-(phenylsulfanyl)propyl]sulfanyl}benzene ≥97%
{[2-Hydroxy-3-(phenylsulfanyl)propyl]sulfanyl}benzene

Hazard: Irritant

Formula: C₁₅H₁₆OS₂
MDL Number: MFCD22126133
CAS Number: 22910-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR