1-(1,1,2,3,3,6-Hexamethyl-2,3-dihydro-1H-inden-5-yl)ethan-1-one ≥95%

Supplier: Matrix Scientific

071348-500MG 071348-1G
102519-836EA 346.15 USD
102519-836 102519-838
1-(1,1,2,3,3,6-Hexamethyl-2,3-dihydro-1H-inden-5-yl)ethan-1-one ≥95%
1-(1,1,2,3,3,6-Hexamethyl-2,3-dihydro-1H-inden-5-yl)ethan-1-one

Hazard: Irritant

Formula: C₁₇H₂₄O
MDL Number: MFCD00451399
CAS Number: 15323-35-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR