3-Fluoro-2-formylpyridine ≥97%
Supplier: Matrix Scientific
Synonyms:
3-Fluoropyridine-2-carboxaldehyde, 3-Fluoropicolinaldehyde, 3-Fluoropyridine-2-carbaldehyde, 3-Fluoro-2-pyridinecarboxaldehyde
071347-1G
071347-5G
102519-834EA
38.46
USD
102519-834
102551-782
3-Fluoro-2-formylpyridine ≥97%
3-Fluoro-2-formylpyridine
Hazard: Irritant
Formula:
C₆H₄FNO MW: 125.1 g/mol Melting Pt: 51-53°C |
MDL Number:
MFCD07781234 CAS Number: 31224-43-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...