5-Chloro-4-(trifluoromethyl)pyridine-2-carboxylic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Chloro-4-(trifluoromethyl)picolinic acid

071333-500MG 071333-1G
102519-776EA 1396.92 USD
102519-776 102519-778
5-Chloro-4-(trifluoromethyl)pyridine-2-carboxylic acid ≥97%
5-Chloro-4-(trifluoromethyl)pyridine-2-carboxylic acid

Hazard: Irritant

Formula: C₇H₃ClF₃NO₂
MW: 225.56 g/mol
Melting Pt: 146…148 °C
MDL Number: MFCD18256834
CAS Number: 796090-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR