(4-Chloro-2-fluoro-5-methoxyphenyl)boronic acid ≥97%

Supplier: Matrix Scientific

071328-500MG 071328-1G
102519-752EA 249.23 USD
102519-752 102519-754
(4-Chloro-2-fluoro-5-methoxyphenyl)boronic acid ≥97%
(4-Chloro-2-fluoro-5-methoxyphenyl)boronic acid

Hazard: Irritant

Formula: C₇H₇BClFO₃
MDL Number: MFCD12912365
CAS Number: 153122-60-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR