5-Bromo-2-(hydroxymethyl)phenol ≥97%

Supplier: Matrix Scientific

071327-500MG 071327-1G
102519-748EA 230.77 USD
102519-748 102519-750
5-Bromo-2-(hydroxymethyl)phenol ≥97%
5-Bromo-2-(hydroxymethyl)phenol

Hazard: Irritant

Formula: C₇H₇BrO₂
Melting Pt: 97…99 °C
MDL Number: MFCD08445665
CAS Number: 170434-11-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR