2-(Methylthio)pyrido[2,3-d]pyrimidin-7(8H)-one ≥97%
Supplier: Matrix Scientific
071260-250MG
071260-1G
102833-496EA
138.46
USD
102833-496
102833-498
2-(Methylthio)pyrido[2,3-d]pyrimidin-7(8H)-one ≥97%
2-(Methylthio)pyrido[2,3-d]pyrimidin-7(8H)-one
Hazard: Irritant
Formula:
C₈H₇N₃OS |
MDL Number:
MFCD03084167 CAS Number: 211244-81-4 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...