2,6-Dibromotoluene ≥97%
Supplier: Matrix Scientific
071179-5G
071179-25G
102519-574EA
56.92
USD
102519-574
102519-576
2,6-Dibromotoluene ≥97%
2,6-Dibromotoluene
Hazard: Irritant
Formula:
C₇H₆Br₂ Boiling Pt: 112-113°C/7mm Melting Pt: 2-6°C Density: 1.812 |
MDL Number:
MFCD00013524 CAS Number: 69321-60-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...