5-Bromothiophene-2-carbaldehyde ≥97%

Supplier: Matrix Scientific

071102-25G 071102-100G
102519-470EA 100 USD
102519-470 102519-472
5-Bromothiophene-2-carbaldehyde ≥97%
5-Bromothiophene-2-carbaldehyde

Hazard: Irritant

Formula: C₅H₃BrOS
Boiling Pt: 104…106 °C (10 mmHg)
Density: 1.762
Flash Pt: 98 °C
MDL Number: MFCD00005432
CAS Number: 4701-17-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR