2-Amino-5-pyridineboronic acid pinacol ester ≥97%

Supplier: Matrix Scientific

Synonyms: 6-Aminopyridine-3-boronic acid pinacol ester, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine, 2-Aminopyridine-5-boronic acid pinacol ester

071082-5G 071082-1G
102519-438EA 229.23 USD
102519-438 102833-486
2-Amino-5-pyridineboronic acid pinacol ester ≥97%
2-Amino-5-pyridineboronic acid pinacol ester

Hazard: Irritant

Formula: C₁₁H₁₇BN₂O₂
MW: 220.08 g/mol
MDL Number: MFCD05663837
CAS Number: 827614-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR