Methyl-3-chlorothiophene-2-carboxylate ≥97%

Supplier: Matrix Scientific

071065-5G 071065-25G
102519-418EA 52.31 USD
102519-418 102519-420
Methyl-3-chlorothiophene-2-carboxylate ≥97%
Methyl-3-chlorothiophene-2-carboxylate

Hazard: Irritant

Formula: C₆H₅ClO₂S
Boiling Pt: 72…74 °C (0.2 mmHg)
Melting Pt: 37…38 °C
Density: 1.28
Flash Pt: 86 °C
MDL Number: MFCD00068135
CAS Number: 88105-17-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR