5-Methylpyridine-2-sulfonamide ≥98%

Supplier: Matrix Scientific

Synonyms: 5-Methyl-2-pyridinesulphonamide

070861-1G 070861-5G
102519-008EA 29.23 USD
102519-008 102519-010
5-Methylpyridine-2-sulfonamide ≥98%
5-Methylpyridine-2-sulfonamide

Hazard: Irritant

Formula: C₆H₈N₂O₂S
MW: 172.21 g/mol
Boiling Pt: 361 °C
Density: 1.346
Flash Pt: 172 °C
MDL Number: MFCD00661363
CAS Number: 65938-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR