11-(2-Chloroacetyl)-5,11-dihydropyrido[2,3-b][1,4]benzodiazepin-6-one ≥98%
Supplier: Matrix Scientific
Synonyms:
11-(2-Chloroacetyl)-5H-benzo[e]pyrido[3,2-b][1,4]diazepin-6(11H)-one
070857-5G
070857-25G
102518-992EA
78.46
USD
102518-992
102518-994
11-(2-Chloroacetyl)-5,11-dihydropyrido[2,3-b][1,4]benzodiazepin-6-one ≥98%
11-(2-Chloroacetyl)-5,11-dihydropyrido[2,3-b][1,4]benzodiazepin-6-one
Hazard: Irritant
Formula:
C₁₄H₁₀ClN₃O₂ MW: 287.703 g/mol Boiling Pt: 481°C Density: 1.415 |
MDL Number:
MFCD21648226 CAS Number: 28797-48-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...