Methyl-2,2-difluorobenzo[d][1,3]dioxole-5-carboxylate ≥98%
Supplier: Matrix Scientific
070856-5G
070856-25G
102518-988EA
73.85
USD
102518-988
102518-990
Methyl-2,2-difluorobenzo[d][1,3]dioxole-5-carboxylate ≥98%
Methyl-2,2-difluorobenzo[d][1,3]dioxole-5-carboxylate
Hazard: Irritant
Formula:
C₉H₆F₂O₄ Density: 1.42 g/cc |
MDL Number:
MFCD06203771 CAS Number: 773873-95-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...