3-Amino-6-bromo-2-methylpyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 5-Amino-2-bromo-6-methylpyridine, 6-Bromo-2-methylpyridin-3-amine

070855-5G 070855-25G
102518-984EA 53.85 USD
102518-984 102518-986
3-Amino-6-bromo-2-methylpyridine ≥98%
3-Amino-6-bromo-2-methylpyridine

Hazard: Irritant

Formula: C₆H₇BrN₂
MW: 187.04 g/mol
MDL Number: MFCD03095086
CAS Number: 126325-47-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR