Methyl-3-bromothiophene-2-carboxylate ≥98%

Supplier: Matrix Scientific

070853-1G 070853-5G
102518-974EA 52.31 USD
102518-974 102518-976
Methyl-3-bromothiophene-2-carboxylate ≥98%
Methyl-3-bromothiophene-2-carboxylate

Hazard: Irritant

Formula: C₆H₅BrO₂S
MDL Number: MFCD00173839
CAS Number: 26137-08-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR