4'-Fluoro-2'-methylacetophenone ≥98%

Supplier: Matrix Scientific

Synonyms: 1-(4-Fluoro-2-methylphenyl)ethanone

070852-1G 070852-5G 070852-25G
102518-968EA 81.54 USD
102518-968 102518-970 102518-972
4'-Fluoro-2'-methylacetophenone ≥98%
4'-Fluoro-2'-methylacetophenone

Hazard: Irritant

Formula: C₉H₉FO
MW: 152.17 g/mol
MDL Number: MFCD00028041
CAS Number: 446-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR