4-Cyano-1-butyne ≥97%

Supplier: Matrix Scientific

070837-1G 070837-5G
102508-804EA 92.31 USD
102508-804 102508-806
4-Cyano-1-butyne ≥97%
4-Cyano-1-butyne

Hazard: Flammable

Formula: C₅H₅N
MW: 79.999 g/mol
Boiling Pt: 60 °C (10 mmHg)
Density: 0.88
Flash Pt: 55 °C
MDL Number: MFCD00041563
CAS Number: 19596-07-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR