Benzyl-3-oxopiperidine-1-carboxylate ≥96%

Supplier: Matrix Scientific

Synonyms: 1-Benzyloxycarbonyl-3-piperidone, Benzyl 3-oxopiperidine-1-carboxylate

070829-1G 070829-5G
102518-944EA 15.38 USD
102518-944 102518-946
Benzyl-3-oxopiperidine-1-carboxylate ≥96%
Benzyl-3-oxopiperidine-1-carboxylate

Hazard: Irritant/Refrigerate

Formula: C₁₃H₁₅NO₃
MW: 233.27 g/mol
Boiling Pt: 384°C
Density: 1.212
MDL Number: MFCD03001723
CAS Number: 61995-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR