Methyl (4-piperidyl)acetate hydrochloride
Supplier: Matrix Scientific
070825-1G
070825-5G
102518-938EA
80
USD
102518-938
102518-940
Methyl (4-piperidyl)acetate hydrochloride
Methyl (4-piperidyl)acetate hydrochloride
Hazard: Irritant
Formula:
C₈H₁₅NO₂ |
MDL Number:
MFCD09261419 CAS Number: 81270-37-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...