4-(4-(Trifluoromethyl)phenyl)pyridine ≥98%

Supplier: Matrix Scientific

070820-1G 070820-5G
102518-932EA 144.62 USD
102518-932 102518-934
4-(4-(Trifluoromethyl)phenyl)pyridine ≥98%
4-(4-(Trifluoromethyl)phenyl)pyridine

Hazard: Irritant

Formula: C₁₂H₈F₃N
MDL Number: MFCD08235161
CAS Number: 220000-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR