1-Bromo-2,6-difluoro-4-nitrobenzene ≥98%

Supplier: Matrix Scientific

070818-1G 070818-5G 070818-25G
102518-924EA 106.15 USD
102518-924 102518-926 102518-928
1-Bromo-2,6-difluoro-4-nitrobenzene ≥98%
1-Bromo-2,6-difluoro-4-nitrobenzene

Hazard: Irritant

Formula: C₆H₂BrF₂NO₂
MDL Number: MFCD04974121
CAS Number: 886762-62-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR