3-Chloro-2-thenoic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Chloro-2-thiophenecarboxylic acid, 3-Chlorothiophene-2-carboxylic acid

070814-5G 070814-25G
102518-920EA 50.77 USD
102518-920 102518-922
3-Chloro-2-thenoic acid ≥98%
3-Chloro-2-thenoic acid

Hazard: Irritant

Formula: C₅H₃ClO₂S
Melting Pt: 185-187°C
MDL Number: MFCD00043888
CAS Number: 59337-89-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR