3-Formyl-N-(tert-butoxycarbonyl)piperidine ≥95%

Supplier: Matrix Scientific

069307-1G 069307-5G 069307-25G
102663-850EA 30.77 USD
102663-850 102663-852 102833-460
3-Formyl-N-(tert-butoxycarbonyl)piperidine ≥95%
3-Formyl-N-(tert-butoxycarbonyl)piperidine

Hazard: Irritant/Air Sensitive/Stored under Argon

Formula: C₁₁H₁₉NO₃
MDL Number: MFCD02179020
CAS Number: 118156-93-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR