N-Boc-3-cyanopyrrolidine ≥95%

Supplier: Matrix Scientific

Synonyms: N-Boc-3-Cyanopyrrolidine

069187-1G 069187-5G
102663-844EA 53.85 USD
102663-844 102663-846
N-Boc-3-cyanopyrrolidine ≥95%
N-Boc-3-cyanopyrrolidine

Hazard: Irritant/Store in Freezer

Formula: C₁₀H₁₆N₂O₂
MW: 196.2462 g/mol
MDL Number: MFCD03001709
CAS Number: 476493-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR