tert-Butyl-N-hydroxycarbamate ≥95%

Supplier: Matrix Scientific

Synonyms: N-Boc-hydroxylamine, N-tert-Butoxycarbonylhydroxylamine, N-Tert-Boc-Hydroxylamine

068961-25G 068961-100G
102551-736EA 29.23 USD
102551-736 102551-738
tert-Butyl-N-hydroxycarbamate ≥95%
tert-Butyl-N-hydroxycarbamate

Hazard: Irritant/Refrigerate

Formula: C₅H₁₁NO₃
Melting Pt: 53…55 °C
MDL Number: MFCD00002107
CAS Number: 36016-38-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR