3-Piperidinemethanol ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(Hydroxymethyl)piperidine

068952-5G 068952-25G
102551-732EA 49.23 USD
102551-732 102551-734
3-Piperidinemethanol ≥95%
3-Piperidinemethanol

Hazard: Corrosive/Hygroscopic

Formula: C₆H₁₃NO
MW: 115.18 g/mol
Boiling Pt: 236 °C
Melting Pt: 58…62 °C
Density: 1.025
MDL Number: MFCD00005997
CAS Number: 4606-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR