Methyl-3-aminobenzoate

Supplier: Matrix Scientific

Synonyms: Methyl 3-aminobenzoate

067795-25G 067795-100G
102551-700EA 113.85 USD
102551-700 102551-702
Methyl-3-aminobenzoate
Methyl-3-aminobenzoate

Hazard: Irritant

Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 152…153 °C (11 Torr)
Melting Pt: 39 °C
MDL Number: MFCD00017102
CAS Number: 4518-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR