2-Bromo-5-methoxyphenol ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Brom-3-hydroxyanisole

067083-1G 067083-5G
102518-908EA 61.54 USD
102518-908 102518-910
2-Bromo-5-methoxyphenol ≥97%
2-Bromo-5-methoxyphenol

Hazard: Irritant

Formula: C₇H₇BrO₂
MDL Number: MFCD03428523
CAS Number: 63604-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR