4-Bromophenethylamine ≥97%

Supplier: Matrix Scientific

Synonyms: 2-(4-Bromophenyl)ethylamine

067077-5G 067077-25G
102833-420EA 92.31 USD
102833-420 102833-422
4-Bromophenethylamine ≥97%
4-Bromophenethylamine

Hazard: Irritant

Formula: C₈H₁₀BrN
MW: 200.08 g/mol
MDL Number: MFCD00008189
CAS Number: 73918-56-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR